3',4'-Dimethoxyacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone
L02737-18
L02737-30
CAAAAL02737-18EA
79.95
CAD
CAAAAL02737-18
CAAAAL02737-30
3',4'-Dimethoxyacetophenone 98+%
3',4'-Dimethoxyacetophenone
Purity: 98+%
CAS No.: 1131-62-0
Molecular Formula: C10H12O3
Formula Weight: 180.20
Flash Point:
Formula:
C₁₀H₁₂O₃ MW: 180.2 g/mol Boiling Pt: 286…288 °C Melting Pt: 47…51 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008737 CAS Number: 1131-62-0 EINECS: 214-468-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...