4-Methylpiperidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02709-AE
L02709-AP
CAAAAL02709-AEEA
58.78
CAD
CAAAAL02709-AE
CAAAAL02709-AP
4-Methylpiperidine 98+%
4-Methylpiperidine
Purity: 98+%
CAS No.: 626-58-4
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 13°(55°F)
Refractive Index: 1.4458
Odor: Amine-like
pH: 13
Melting Point: 4-5°
Boiling Point: 123-124°
Beilstein Reference: 1083
UN No.: 2733
MDL No.: MFCD00006005
Formula:
C₆H₁₃N MW: 99.18 g/mol Boiling Pt: 123…124 °C Melting Pt: 4…5 °C Density: 0.838 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006005 CAS Number: 626-58-4 EINECS: 210-954-1 UN: 3286 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...