3-Hexyne 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02682-06
L02682-14
CAAAAL02682-06EA
58.78
CAD
CAAAAL02682-06
CAAAAL02682-14
3-Hexyne 99%
3-Hexyne
Purity: 99%
CAS No.: 928-49-4
Molecular Formula: C6H10
Formula Weight: 82.15
Flash Point: -10°(14°F)
Refractive Index: 1.4110
Odor: Characteristic
Boiling Point: 80-82°
Beilstein Reference: 1731158
UN No.: 3295
MDL No.: MFCD00009381
Formula:
C₆H₁₀ Boiling Pt: 80…82 °C Melting Pt: –61.5 °C Density: 0.721 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009381 CAS Number: 928-49-4 EINECS: 213-173-4 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...