3-Iodopropionic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02655-06
L02655-14
CAAAAL02655-06EA
52.93
CAD
CAAAAL02655-06
CAAAAL02655-14
3-Iodopropionic acid 99%
3-Iodopropionic acid
Purity: 99%
CAS No.: 141-76-4
Molecular Formula: ICH2CH2CO2H
Formula Weight: 199.98
Melting Point: 80-83°
Beilstein Reference: 1699514
UN No.: 3261
MDL No.: MFCD00002765
Formula:
C₃H₅IO₂ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002765 CAS Number: 141-76-4 EINECS: 205-499-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...