1-Bromo-4-methylcyclohexane cis- and trans-mixture 97%

Supplier: Thermo Scientific Chemicals

L02640-06 L02640-14
CAAAAL02640-06EA 122.77 CAD
CAAAAL02640-06 CAAAAL02640-14
1-Bromo-4-methylcyclohexane cis- and trans-mixture 97%
1-Bromo-4-methylcyclohexane

Purity: 97% CAS No.: 6294-40-2 Molecular Formula: C7H13Br Formula Weight: 177.09 Refractive Index: 1.4880 Boiling Point: 94-96°/50mm MDL No.: MFCD00014282

Formula: C₇H₁₃Br
Boiling Pt: 94…96 °C/50 mm
Density: 1.262
Storage Temperature: Ambient
MDL Number: MFCD00014282
CAS Number: 6294-40-2
EINECS: 228-559-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR