Benzyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02633-03
L02633-06
L02633-14
CAAAAL02633-03EA
45.35
CAD
CAAAAL02633-03
CAAAAL02633-06
CAAAAL02633-14
Benzyl isocyanate 98%
Benzyl isocyanate
Purity: 98%
CAS No.: 3173-56-6
Molecular Formula: C8H7NO
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Flash Point: 43°(109°F)
Refractive Index: 1.5250
Odor: Pungent
Boiling Point: 53-54°/0.8mm
Beilstein Reference: 636508
UN No.: 3080
MDL No.: MFCD00009701
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 53…54 °C/0.8 mm Density: 1.078 Flash Pt: 43 °C (109 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009701 CAS Number: 3173-56-6 EINECS: 221-640-9 UN: 3080 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...