5-Cyano-1-pentyne 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02604-03
L02604-06
CAAAAL02604-03EA
56.72
CAD
CAAAAL02604-03
CAAAAL02604-06
5-Cyano-1-pentyne 98%
5-Cyano-1-pentyne
Purity: 98%
CAS No.: 14918-21-9
Molecular Formula: HC≡CCH2CH2CH2CN
Formula Weight: 93.13
Storage Temperature: 2°C to 8°C
Flash Point: 42°(107°F)
Refractive Index: 1.4410
Boiling Point: 115-117°
Beilstein Reference: 1735926
UN No.: 1992
MDL No.: MFCD00001978
Formula:
C₆H₇N Boiling Pt: 115…117 °C Density: 0.889 Flash Pt: 42 °C (107 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001978 CAS Number: 14918-21-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...