1-Tetradecyne tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

L02573-06 L02573-14
CAAAAL02573-06EA 103.65 CAD
CAAAAL02573-06 CAAAAL02573-14
1-Tetradecyne tech. 90%, Technical Grade
1-Tetradecyne

Purity: 90% CAS No.: 765-10-6 Molecular Formula: CH3(CH2)11C≡CH Formula Weight: 194.36 Refractive Index: 1.4410 Boiling Point: 132-134°/14mm MDL No.: MFCD00015076

Formula: C₁₄H₂₆
Boiling Pt: 132…134 °C/14 mm
Density: 0.790
Storage Temperature: Ambient
MDL Number: MFCD00015076
CAS Number: 765-10-6
EINECS: 212-139-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR