2-Methylpiperidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02567-AE
L02567-AP
CAAAAL02567-AEEA
62.88
CAD
CAAAAL02567-AE
CAAAAL02567-AP
2-Methylpiperidine 98+%
2-Methylpiperidine
Purity: 98+%
CAS No.: 109-05-7
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 8°(46°F)
Refractive Index: 1.4470
Odor: Amine-like
Melting Point: -4°
Boiling Point: 118-119°
Beilstein Reference: 79804
UN No.: 2733
MDL No.: MFCD00005982
Formula:
C₆H₁₃N MW: 99.18 g/mol Boiling Pt: 118…119 °C Melting Pt: –4 °C Density: 0.842 Flash Pt: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005982 CAS Number: 109-05-7 EINECS: 203-642-1 UN: 2733 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...