3-(2-Hydroxyethyl)indole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol

L02555-03 L02555-06
CAAAAL02555-03EA 44.87 CAD
CAAAAL02555-03 CAAAAL02555-06
3-(2-Hydroxyethyl)indole 97%
3-(2-Hydroxyethyl)indole

Purity: 97% CAS No.: 526-55-6 Molecular Formula: C10H11NO Formula Weight: 161.20 Flash Point: 255°(491°F) Melting Point: 56-59° Boiling Point: 174°/2mm Beilstein Reference: 125553 Merck Reference: 14,9798 MDL No.: MFCD00005659

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 358
Melting Pt: 56…59 °C
Flash Pt: 255 °C (491 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005659
CAS Number: 526-55-6
EINECS: 208-393-2
Merck Index: 14,9798

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR