Triphenylphosphine 99%, powder

Supplier: Thermo Scientific Chemicals
Warning

L02502-0B L02502-18 L02502-30
CAAAAL02502-0BEA 440.82 CAD
CAAAAL02502-0B CAAAAL02502-18 CAAAAL02502-30
Triphenylphosphine 99%, powder
Triphenylphosphine

Purity: 99% CAS No.: 603-35-0 Molecular Formula: (C6H5)3P Formula Weight: 262.29 Flash Point: 181°(358°F) Odor: Mild Boiling Point: 360° Beilstein Reference: 610776 Merck Reference: 14,9743 MDL No.: MFCD00003043

Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377
Melting Pt: 79
Density: 1.200
Flash Pt: 181 °C (358 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 14,9743

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR