1-Pentyne 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02479-06
L02479-14
CAAAAL02479-06EA
49.45
CAD
CAAAAL02479-06
CAAAAL02479-14
1-Pentyne 99%
1-Pentyne
Purity: 99%
CAS No.: 627-19-0
Molecular Formula: C5H8
Formula Weight: 68.12
Storage Temperature: 2°C to 8°C
Flash Point: -20°(-4°F)
Refractive Index: 1.3860
Odor: Unpleasant
Melting Point: -106°
Boiling Point: 39-41°
Beilstein Reference: 1697133
UN No.: 3295
MDL No.: MFCD00009469
Formula:
C₅H₈ Boiling Pt: 39…41 °C Melting Pt: –106 °C Density: 0.692 Flash Pt: –20 °C (–4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009469 CAS Number: 627-19-0 EINECS: 210-987-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...