4-tert-Butylcyclohexylamine cis- and trans-mixture 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(tert-Butyl)cyclohexanamine

L02472-06 L02472-14
CAAAAL02472-06EA 75.52 CAD
CAAAAL02472-06 CAAAAL02472-14
4-tert-Butylcyclohexylamine cis- and trans-mixture 97%
4-tert-Butylcyclohexylamine

Purity: 97% CAS No.: 5400-88-4 Molecular Formula: C10H21N Formula Weight: 155.29 Flash Point: 79°(174°F) Refractive Index: 1.4662 Beilstein Reference: 1523780 UN No.: 2735 MDL No.: MFCD00013763

Formula: C₁₀H₂₁N
MW: 155.28 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.870
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013763
CAS Number: 5400-88-4
EINECS: 226-444-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR