2-Methylnaphthalene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
β-Methylnaphthalene
L02459-22
L02459-36
CAAAAL02459-22EA
44.24
CAD
CAAAAL02459-22
CAAAAL02459-36
2-Methylnaphthalene 97%
2-Methylnaphthalene
Purity: 97%
CAS No.: 91-57-6
Molecular Formula: C11H10
Formula Weight: 142.20
Flash Point: 97°(206°F)
Odor: Characteristic
Boiling Point: 241-243°
Beilstein Reference: 906859
UN No.: 3077
MDL No.: MFCD00004118
Formula:
C₁₁H₁₀ MW: 142.2 g/mol Boiling Pt: 241…243 °C Melting Pt: 31…35 °C Density: 1.001 Flash Pt: 97 °C(206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004118 CAS Number: 91-57-6 EINECS: 202-078-3 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...