Bromopentamethylbenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02451-06
L02451-14
CAAAAL02451-06EA
58.46
CAD
CAAAAL02451-06
CAAAAL02451-14
Bromopentamethylbenzene 98+%
Bromopentamethylbenzene
Purity: 98+%
CAS No.: 5153-40-2
Molecular Formula: C11H15Br
Formula Weight: 227.15
Melting Point: 161-162°
Beilstein Reference: 1863555
Formula:
C₁₁H₁₅Br Melting Pt: 161…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013523 CAS Number: 5153-40-2 EINECS: 225-926-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...