2-Propanesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isopropylsulphonyl chloride, Isopropylsulfonyl chloride
L02431-06
L02431-14
CAAAAL02431-06EA
73.63
CAD
CAAAAL02431-06
CAAAAL02431-14
2-Propanesulfonyl chloride 97%
2-Propanesulphonyl chloride
Purity: 97%
CAS No.: 10147-37-2
Molecular Formula: (CH3)2CHSO2Cl
Formula Weight: 142.60
Flash Point: 86°(186°F)
Refractive Index: 1.4560
Melting Point: -47°
Boiling Point: 95°/45mm
Beilstein Reference: 1747497
UN No.: 3265
MDL No.: MFCD00007453
Formula:
C₃H₇ClO₂S MW: 142.61 g/mol Boiling Pt: 95 °C/45 mm Melting Pt: –47 °C Density: 1.273 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007453 CAS Number: 10147-37-2 EINECS: 233-415-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...