(1-Octyl)triphenylphosphonium bromide 97%

Supplier: Thermo Scientific Chemicals
Warning

L02412-06 L02412-14 L02412-22
CAAAAL02412-06EA 92.59 CAD
CAAAAL02412-06 CAAAAL02412-14 CAAAAL02412-22
(1-Octyl)triphenylphosphonium bromide 97%
(1-Octyl)triphenylphosphonium bromide

Purity: 97% CAS No.: 42036-78-2 Molecular Formula: C26H32BrP Formula Weight: 455.42 Melting Point: 61-63° Beilstein Reference: 3642460

Formula: H₃C(CH₂)₇P(C₆H₅)₃Br
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00051874
CAS Number: 42036-78-2
EINECS: 255-635-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d20265a4fb0cff86d083b39bfedb8c80cf4883dc&vid=d20265a4fb0cff86d083b39bfedb8c80cf4883dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR