Diphenylacetonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02404-22
L02404-36
CAAAAL02404-22EA
52.77
CAD
CAAAAL02404-22
CAAAAL02404-36
Diphenylacetonitrile 99%
Diphenylacetonitrile
Purity: 99%
CAS No.: 86-29-3
Molecular Formula: C14H11N
Formula Weight: 193.25
Flash Point: 120°(248°F)
Melting Point: 72-76°
Boiling Point: 181°/12mm
Beilstein Reference: 1911160
Merck Reference: 14,3316
MDL No.: MFCD00001862
Formula:
(C₆H₅)₂CHCN Boiling Pt: 181 °C/12 mm Melting Pt: 72…76 °C Density: 1.182 g/cm³ (20 °C) Flash Pt: 120 °C(248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001862 CAS Number: 86-29-3 EINECS: 201-662-5 Merck Index: 14,3316 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...