1-Chloro-2,2-dimethylpropane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride

L02365-06 L02365-14
CAAAAL02365-06EA 80.42 CAD
CAAAAL02365-06 CAAAAL02365-14
1-Chloro-2,2-dimethylpropane 98%
1-Chloro-2,2-dimethylpropane

Purity: 98% CAS No.: 753-89-9 Molecular Formula: C5H11Cl Formula Weight: 106.60 Flash Point: -8°(17°F) Refractive Index: 1.4039 Odor: Sweet Melting Point: -20° Boiling Point: 83-84° Beilstein Reference: 1730985 UN No.: 1107 MDL No.: MFCD00000941

Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 84
Melting Pt: -20
Density: 0.868
Flash Pt: –8 °C (17 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000941
CAS Number: 753-89-9
EINECS: 212-040-8
UN: 1107
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR