2-Methoxybenzylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02273-14
L02273-22
CAAAAL02273-14EA
147.26
CAD
CAAAAL02273-14
CAAAAL02273-22
2-Methoxybenzylamine 98+%
2-Methoxybenzylamine
Purity: 98+%
CAS No.: 6850-57-3
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 100°(212°F)
Refractive Index: 1.5480
Boiling Point: 228-229°
Beilstein Reference: 1100448
UN No.: 2735
MDL No.: MFCD00008110
Formula:
C₈H₁₁NO Boiling Pt: 228…229 °C Density: 1.057 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008110 CAS Number: 6850-57-3 EINECS: 229-941-7 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...