α-Phenylbenzyl bromide 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02211-14
L02211-22
L02211-36
CAAAAL02211-14EA
47.87
CAD
CAAAAL02211-14
CAAAAL02211-22
CAAAAL02211-36
α-Phenylbenzyl bromide 90+%
α-Phenylbenzyl bromide
Purity: 90+%
CAS No.: 776-74-9
Molecular Formula: C13H11Br
Formula Weight: 247.14
Flash Point:
Formula:
(C₆H₅)₂CHBr Boiling Pt: 183…185 °C/20 mm Melting Pt: 36…41 °C Density: 1.491 g/cm³ (13 °C) Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000134 CAS Number: 776-74-9 EINECS: 212-279-8 UN: 1770 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...