N,N'-Dimethylethylendiamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine
L02204-06
L02204-14
CAAAAL02204-06EA
94.96
CAD
CAAAAL02204-06
CAAAAL02204-14
N,N'-Dimethylethylendiamine 95%
N,N'-Dimethylethylendiamine
Purity: 95%
CAS No.: 110-70-3
Molecular Formula: C4H12N2
Formula Weight: 88.15
Flash Point: 28°(82°F)
Refractive Index: 1.4300
Odor: Amine-like
Boiling Point: 118-119°
Beilstein Reference: 878142
UN No.: 2734
MDL No.: MFCD00008290
Formula:
C₄H₁₂N₂ Boiling Pt: 118…119 °C Density: 0.818 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008290 CAS Number: 110-70-3 EINECS: 203-793-3 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...