Acetylcholine chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02168-14
L02168-22
CAAAAL02168-14EA
96.06
CAD
CAAAAL02168-14
CAAAAL02168-22
Acetylcholine chloride 98+%
Acetylcholine chloride
Endogenous neurotransmitter at cholinergic synapses
Purity: 98+%
CAS No.: 60-31-1
Molecular Formula: CH3CO2CH2CH2N(CH3)3Cl
Formula Weight: 181.66
Storage Temperature: 2°C to 8°C
Melting Point: 149-152°
Beilstein Reference: 3571875
Merck Reference: 14,87
MDL No.: MFCD00011698
Formula:
H₃CC(O)O(CH₂)₂N(CH₃)₃Cl Melting Pt: 149…152 °C Density: 1.263 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011698 CAS Number: 60-31-1 EINECS: 200-468-8 Merck Index: 14,87 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...