2,4,6-Trimethoxytoluene 97%

Supplier: Thermo Scientific Chemicals

L02155-06 L02155-14
CAAAAL02155-06EA 240.16 CAD
CAAAAL02155-06 CAAAAL02155-14
2,4,6-Trimethoxytoluene 97%
2,4,6-Trimethoxytoluene

Purity: 97% CAS No.: 14107-97-2 Molecular Formula: C10H14O3 Formula Weight: 182.22 Refractive Index: 1.5310 Melting Point: 27-28° Boiling Point: 100-105°/1mm Beilstein Reference: 2048952 MDL No.: MFCD00008375

Formula: C₁₀H₁₄O₃
Boiling Pt: 100…105 °C/1 mm
Melting Pt: 27…28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008375
CAS Number: 14107-97-2
EINECS: 237-962-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR