p-Ethyltoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02120-06
L02120-14
L02120-22
CAAAAL02120-06EA
61.94
CAD
CAAAAL02120-06
CAAAAL02120-14
CAAAAL02120-22
p-Ethyltoluene 97%
p-Ethyltoluene
Purity: 97%
CAS No.: 622-96-8
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 43°(109°F)
Refractive Index: 1.4950
Melting Point: -62°
Boiling Point: 161-163°
Beilstein Reference: 1903007
UN No.: 3295
MDL No.: MFCD00009263
Formula:
C₉H₁₂ Boiling Pt: 161…163 °C Melting Pt: –62 °C Density: 0.861 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009263 CAS Number: 622-96-8 EINECS: 210-761-2 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...