2-Aminobenzimidazole 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole
L02066-06
L02066-14
L02066-22
CAAAAL02066-06EA
55.3
CAD
CAAAAL02066-06
CAAAAL02066-14
CAAAAL02066-22
2-Aminobenzimidazole 97+%
2-Aminobenzimidazole
Purity: 97+%
CAS No.: 934-32-7
Molecular Formula: C7H7N3
Formula Weight: 133.15
Melting Point: 228-232°
Beilstein Reference: 116525
Formula:
C₇H₇N₃ MW: 133.15 g/mol Boiling Pt: 381 °C (1013 hPa) Melting Pt: 228…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005596 CAS Number: 934-32-7 EINECS: 213-280-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...