3-Methoxybenzylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

L02054-06 L02054-14
CAAAAL02054-06EA 87.06 CAD
CAAAAL02054-06 CAAAAL02054-14
3-Methoxybenzylamine 98+%
3-Methoxybenzylamine

Purity: 98+% CAS No.: 5071-96-5 Molecular Formula: C8H11NO Formula Weight: 137.18 Flash Point: 110°(230°F) Refractive Index: 1.5470 Boiling Point: 230-231° Beilstein Reference: 1447182 UN No.: 2735 MDL No.: MFCD00008115

Formula: C₈H₁₁NO
Boiling Pt: 230…231 °C
Density: 1.066
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR