o-Ethyltoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02028-06
L02028-14
CAAAAL02028-06EA
86.58
CAD
CAAAAL02028-06
CAAAAL02028-14
o-Ethyltoluene 98+%
o-Ethyltoluene
Purity: 98+%
CAS No.: 611-14-3
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 53°(127°F)
Refractive Index: 1.5040
Melting Point: -17°
Boiling Point: 164-165°
Beilstein Reference: 1851237
UN No.: 3295
MDL No.: MFCD00009257
Formula:
C₉H₁₂ Boiling Pt: 164…165 °C Melting Pt: –17 °C Density: 0.884 Flash Pt: 53 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009257 CAS Number: 611-14-3 EINECS: 210-255-1 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...