4,5-Dibromoveratrole 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2-Dibromo-4,5-dimethoxybenzene, 4,5-Dibromoveratrole

L01999-09 L01999-18 L01999-30
CAAAAL01999-09EA 85.48 CAD
CAAAAL01999-09 CAAAAL01999-18 CAAAAL01999-30
4,5-Dibromoveratrole 98+%
4,5-Dibromoveratrole

Purity: 98+% CAS No.: 37895-73-1 Molecular Formula: C8H8Br2O2 Formula Weight: 295.96 Melting Point: 90-92° Beilstein Reference: 1953148

Formula: C₈H₈Br₂O₂
MW: 295.96 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00014894
CAS Number: 37895-73-1
EINECS: 253-705-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR