2,4,6-Trimethylbenzyl alcohol 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: (2,4,6-Trimethylphenyl)methanol, Mesitylmethanol

L01952-06 L01952-14
CAAAAL01952-06EA 104.28 CAD
CAAAAL01952-06 CAAAAL01952-14
2,4,6-Trimethylbenzyl alcohol 98+%
2,4,6-Trimethylbenzyl alcohol

Purity: 98+% CAS No.: 4170-90-5 Molecular Formula: C10H14O Formula Weight: 150.22 Melting Point: 85-89° Boiling Point: 140°/15mm MDL No.: MFCD00014422

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 140 °C/15 mm
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00014422
CAS Number: 4170-90-5
EINECS: 224-032-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR