3-Thiophenemethanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01935-06
L01935-14
CAAAAL01935-06EA
165.9
CAD
CAAAAL01935-06
CAAAAL01935-14
3-Thiophenemethanol 98%
3-Thiophenemethanol
Purity: 98%
CAS No.: 71637-34-8
Molecular Formula: C5H6OS
Formula Weight: 114.17
Flash Point: 110°(230°F)
Refractive Index: 1.5650
Boiling Point: 86-88°/10mm
Beilstein Reference: 106460
MDL No.: MFCD00014534
Formula:
C₅H₆OS Boiling Pt: 86…88 °C/10 mm Density: 1.210 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014534 CAS Number: 71637-34-8 EINECS: 275-741-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...