(±)-2-Tridecanol 97%
Supplier: Thermo Scientific Chemicals
L01926-03
L01926-06
CAAAAL01926-03EA
45.82
CAD
CAAAAL01926-03
CAAAAL01926-06
(±)-2-Tridecanol 97%
(±)-2-Tridecanol
Purity: 97%
CAS No.: 1653-31-2
Molecular Formula: C13H28O
Formula Weight: 200.37
Refractive Index: 1.4430
Melting Point: 23-26°
Boiling Point: 265°
Beilstein Reference: 1720217
MDL No.: MFCD00037601
Formula:
C₁₃H₂₈O Boiling Pt: 265 °C Melting Pt: 23…26 °C Density: 0,83 Storage Temperature: Ambient |
MDL Number:
MFCD00037601 CAS Number: 1653-31-2 EINECS: 216-723-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...