2,5-Dimethylbenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L01924-06
L01924-14
CAAAAL01924-06EA
115.5
CAD
CAAAAL01924-06
CAAAAL01924-14
2,5-Dimethylbenzaldehyde 98%
2,5-Dimethylbenzaldehyde
Purity: 98%
CAS No.: 5779-94-2
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 87°(188°F)
Refractive Index: 1.5440
Odor: Almond-like
Boiling Point: 104-106°/14mm
Beilstein Reference: 1927303
MDL No.: MFCD00003342
Formula:
C₉H₁₀O Boiling Pt: 104…106 °C/14 mm Density: 0.950 Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003342 CAS Number: 5779-94-2 EINECS: 227-303-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...