Phenanthrene 98%

Supplier: Thermo Scientific Chemicals
Warning

L01921-18 L01921-30
CAAAAL01921-18EA 134.62 CAD
CAAAAL01921-18 CAAAAL01921-30
Phenanthrene 98%
Phenanthrene

Purity: 98% CAS No.: 85-01-8 Molecular Formula: C14H10 Formula Weight: 178.23 Flash Point: 171°(339°F) Melting Point: 97-101° Boiling Point: 340° Beilstein Reference: 1905428 Merck Reference: 14,7212 UN No.: 3077 MDL No.: MFCD00001168

Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340
Melting Pt: 97…101 °C
Density: 1.179
Flash Pt: 171 °C (339 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9 III
Merck Index: 14,7212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92527d856e1fe8582e54bf3812e1580ee0b91b05&vid=d8ec397dac001e49a7958f12aacb577cd11954da&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR