1-Bromo-1-chloroethane 98%

Supplier: Thermo Scientific Chemicals
Warning

L01910-04 L01910-09
CAAAAL01910-04EA 152.94 CAD
CAAAAL01910-04 CAAAAL01910-09
1-Bromo-1-chloroethane 98%
1-Bromo-1-chloroethane

Purity: 98% CAS No.: 593-96-4 Molecular Formula: CH3CHBrCl Formula Weight: 143.42 Refractive Index: 1.4660 Boiling Point: 82-83° UN No.: 2810 MDL No.: MFCD00018842

Formula: BrClHCCH₃
Boiling Pt: 82…83 °C
Density: 1.667
Storage Temperature: Ambient
MDL Number: MFCD00018842
CAS Number: 593-96-4
EINECS: 209-820-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR