6-Nitro-o-toluidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Nitro-2-methylaniline, 2-Amino-3-nitrotoluene, 2-Methyl-6-nitroaniline

L01803-14 L01803-22
CAAAAL01803-14EA 175.38 CAD
CAAAAL01803-14 CAAAAL01803-22
6-Nitro-o-toluidine 98%
6-Nitro-o-toluidine

Purity: 98% CAS No.: 570-24-1 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: 93-96° Beilstein Reference: 1868029 UN No.: 2660 MDL No.: MFCD00007744

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 120…124 °C (1 mmHg)
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007744
CAS Number: 570-24-1
EINECS: 209-329-6
UN: 2660
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR