2,4-Pentanediol (±) + meso 99%
Supplier: Thermo Scientific Chemicals
L01779-09
L01779-18
CAAAAL01779-09EA
120.71
CAD
CAAAAL01779-09
CAAAAL01779-18
2,4-Pentanediol (±) + meso 99%
2,4-Pentanediol
Purity: 99%
CAS No.: 625-69-4
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 101°(213°F)
Refractive Index: 1.4350
Odor: Odorless
Boiling Point: 200-201°
Beilstein Reference: 969186
MDL No.: MFCD00004549
Formula:
C₅H₁₂O₂ Boiling Pt: 200…201 °C Melting Pt: ∼53 °C Density: 0.955 Flash Pt: 101 °C(213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004549 CAS Number: 625-69-4 EINECS: 210-907-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...