2,5-Dibromo-p-xylene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Dibromo-1,4-dimethylbenzene, 1,4-Dibromo-2,5-dimethylbenzene

L01776-14 L01776-22
CAAAAL01776-14EA 75.52 CAD
CAAAAL01776-14 CAAAAL01776-22
2,5-Dibromo-p-xylene 98+%
2,5-Dibromo-p-xylene

Purity: 98+% CAS No.: 1074-24-4 Molecular Formula: C8H8Br2 Formula Weight: 263.97 Odor: Odorless Melting Point: 72-74° Boiling Point: 261° Beilstein Reference: 2080433 MDL No.: MFCD00000091

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 261 °C
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00000091
CAS Number: 1074-24-4
EINECS: 214-038-2
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR