1,1,1,5,5,5-Hexafluoroacetylacetone 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA
L01693-06
L01693-14
L01693-22
CAAAAL01693-06EA
91.01
CAD
CAAAAL01693-06
CAAAAL01693-14
CAAAAL01693-22
1,1,1,5,5,5-Hexafluoroacetylacetone 98+%
1,1,1,5,5,5-Hexafluoroacetylacetone
Purity: 98+%
CAS No.: 1522-22-1
Molecular Formula: C5H2F6O2
Formula Weight: 208.06
Flash Point: 32°(89°F)
Refractive Index: 1.3340
Boiling Point: 69-71°
Beilstein Reference: 1790138
UN No.: 2920
MDL No.: MFCD00000426
Formula:
C₅H₂F₆O₂ MW: 208.06 g/mol Boiling Pt: 69…71 °C Density: 1.486 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000426 CAS Number: 1522-22-1 EINECS: 216-191-0 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...