3-Methoxysalicylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxy-3-methoxybenzoic acid
L01658-06
L01658-14
CAAAAL01658-06EA
39.82
CAD
CAAAAL01658-06
CAAAAL01658-14
3-Methoxysalicylic acid 98+%
3-Methoxysalicylic acid
Purity: 98+%
CAS No.: 877-22-5
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 151-152°
Beilstein Reference: 2209642
Formula:
C₈H₈O₄ Melting Pt: 151…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002445 CAS Number: 877-22-5 EINECS: 212-888-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...