9-Heptadecanone 98%

Supplier: Thermo Scientific Chemicals

L01630-06 L01630-14
CAAAAL01630-06EA 110.44 CAD
CAAAAL01630-06 CAAAAL01630-14
9-Heptadecanone 98%
9-Heptadecanone

Purity: 98% CAS No.: 540-08-9 Molecular Formula: C17H34O Formula Weight: 254.46 Melting Point: 51-53° Boiling Point: 250-253° MDL No.: MFCD00009573

Formula: C₁₇H₃₄O
MW: 254.46 g/mol
Boiling Pt: 250…253 °C
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00009573
CAS Number: 540-08-9
EINECS: 208-734-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR