2,5-Dimethylbenzyl alcohol 97+%
Supplier: Thermo Scientific Chemicals
L01624-03
L01624-06
CAAAAL01624-03EA
87.69
CAD
CAAAAL01624-03
CAAAAL01624-06
2,5-Dimethylbenzyl alcohol 97+%
2,5-Dimethylbenzyl alcohol
Purity: 97+%
CAS No.: 53957-33-8
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 107°(224°F)
Refractive Index: 1.5360
Melting Point: 40-42°
Boiling Point: 232-234°
Beilstein Reference: 2243420
MDL No.: MFCD00004625
Formula:
C₉H₁₂O Boiling Pt: 232…234 °C Melting Pt: 40…42 °C Density: 1,011 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004625 CAS Number: 53957-33-8 EINECS: 258-888-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...