3,4,5-Trimethoxyaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01619-06
L01619-14
CAAAAL01619-06EA
79
CAD
CAAAAL01619-06
CAAAAL01619-14
3,4,5-Trimethoxyaniline 98+%
3,4,5-Trimethoxyaniline
Purity: 98+%
CAS No.: 24313-88-0
Molecular Formula: C9H13NO3
Formula Weight: 183.21
Melting Point: 110-113°
Beilstein Reference: 642919
UN No.: 2811
MDL No.: MFCD00008393
Formula:
C₉H₁₃NO₃ Boiling Pt: 293 °C (1013 hPa) Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008393 CAS Number: 24313-88-0 EINECS: 246-154-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...