2-Fluoro-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-4-picoline
L01602-03
L01602-06
CAAAAL01602-03EA
49.77
CAD
CAAAAL01602-03
CAAAAL01602-06
2-Fluoro-4-methylpyridine 98%
2-Fluoro-4-methylpyridine
Purity: 98%
CAS No.: 461-87-0
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 51°(123°F)
Refractive Index: 1.4725
Boiling Point: 155-156°
Beilstein Reference: 107086
UN No.: 1993
MDL No.: MFCD00041224
Formula:
C₆H₆FN MW: 111.12 g/mol Boiling Pt: 157 Density: 1.078 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041224 CAS Number: 461-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...