1,2-Dibenzoylbenzene 97%
Supplier: Thermo Scientific Chemicals
L01592-03
L01592-MD
CAAAAL01592-03EA
156.1
CAD
CAAAAL01592-03
CAAAAL01592-MD
1,2-Dibenzoylbenzene 97%
1,2-Dibenzoylbenzene
Purity: 97%
CAS No.: 1159-86-0
Molecular Formula: C20H14O2
Formula Weight: 286.33
Melting Point: 145-148°
Beilstein Reference: 789077
Formula:
C₂₀H₁₄O₂ Boiling Pt: 424 °C (1013 hPa) Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003078 CAS Number: 1159-86-0 EINECS: 214-597-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...