2,4-Dichlorophenylboronic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
L01563-03
L01563-06
CAAAAL01563-03EA
39.18
CAD
CAAAAL01563-03
CAAAAL01563-06
2,4-Dichlorophenylboronic acid 98+%
2,4-Dichlorophenylboronic acid
Purity: 98+%
CAS No.: 68716-47-2
Molecular Formula: C6H5BCl2O2
Formula Weight: 190.82
Melting Point: 242-245°
Beilstein Reference: 4983878
Formula:
C₆H₅BCl₂O₂ MW: 190.82 g/mol Melting Pt: 242…245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013930 CAS Number: 68716-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...