2-Fluoro-6-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-6-picoline

L01561-03 L01561-06
CAAAAL01561-03EA 29.55 CAD
CAAAAL01561-03 CAAAAL01561-06
2-Fluoro-6-methylpyridine 98%
2-Fluoro-6-methylpyridine

Purity: 98% CAS No.: 407-22-7 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 66°(150°F) Refractive Index: 1.4700 Boiling Point: 140-141° Beilstein Reference: 107084 MDL No.: MFCD00041226

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 140…141 °C
Density: 1.077
Flash Pt: 66 °C(150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041226
CAS Number: 407-22-7
EINECS: 206-980-8
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR