3,4,5-Trimethoxyphenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01527-06
L01527-14
CAAAAL01527-06EA
118.18
CAD
CAAAAL01527-06
CAAAAL01527-14
3,4,5-Trimethoxyphenol 97%
3,4,5-Trimethoxyphenol
Purity: 97%
CAS No.: 642-71-7
Molecular Formula: C9H12O4
Formula Weight: 184.19
Melting Point: 143-148°
Beilstein Reference: 1958573
Formula:
C₉H₁₂O₄ Boiling Pt: 283 °C (1013 hPa) Melting Pt: 143…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008389 CAS Number: 642-71-7 EINECS: 211-387-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...