3-Fluorophenyl isocyanate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L01496-06
L01496-14
CAAAAL01496-06EA
101.91
CAD
CAAAAL01496-06
CAAAAL01496-14
3-Fluorophenyl isocyanate 97+%
3-Fluorophenyl isocyanate
Purity: 97+%
CAS No.: 404-71-7
Molecular Formula: C7H4FNO
Formula Weight: 137.11
Storage Temperature: 2°C to 8°C
Flash Point: 41°(105°F)
Refractive Index: 1.5141
Odor: Pungent, makes eyes water
Boiling Point: 56-58°/14mm
Beilstein Reference: 774879
UN No.: 3080
MDL No.: MFCD00002015
Formula:
C₇H₄FNO Boiling Pt: 56…58 °C/14 mm Density: 1.207 Flash Pt: 41 °C (105 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002015 CAS Number: 404-71-7 EINECS: 206-966-1 UN: 3080 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...