3-Bromo-1,2,4,5-tetramethylbenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01465-06
L01465-14
CAAAAL01465-03EA
42.57
CAD
CAAAAL01465-03
CAAAAL01465-06
CAAAAL01465-14
3-Bromo-1,2,4,5-tetramethylbenzene 99%
3-Bromo-1,2,4,5-tetramethylbenzene
Purity: 99%
CAS No.: 1646-53-3
Molecular Formula: C10H13Br
Formula Weight: 213.12
Melting Point: 58-62°
Boiling Point: 115-120°/5mm
MDL No.: MFCD00000070
Formula:
C₁₀H₁₃Br Boiling Pt: 115…120 °C/5 mm Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000070 CAS Number: 1646-53-3 EINECS: 216-707-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...